λ DNA (cIind 1 ts857 Sam 7) is isolated from a dam+ dcm+ E. coli LE597 lysogen. The DNA isRead more
Have Questions?
Catalog Number
Quantity
25250010
500 μg
Catalog number 25250010
Price (INR)
12,513.00
Online offer
Ends: 31-Mar-2026
20,855.00
Save 8,342.00 (40%)
Each
Quantity:
500 μg
Price (INR)
12,513.00
Online offer
Ends: 31-Mar-2026
20,855.00
Save 8,342.00 (40%)
Each
λ DNA (cIind 1 ts857 Sam 7) is isolated from a dam+ dcm+ E. coli LE597 lysogen. The DNA is suitable for use as a substrate for generating molecular size standards and for testing restriction endonuclease activity. Restriction endonucleases sensitive to dam or dcm methylation will not cleave this DNA.
Performance and Quality Testing: Purity, DNA structure, and selected restriction endonuclease fragmentation patterns are verified by agarose gel analysis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
IncludesControl Template(s)
Label or DyeUnconjugated
Product Typeλ DNA
Quantity500 μg
Shipping ConditionWet Ice
FormatTube
Unit SizeEach
Contents & Storage
Supplied at 0.25 to 0.6 μg/μl in 10 mM Tris-HCl (pH 7.4), 5 mM NaCl, 0.1 mM EDTA. Store at +4°C.