Sodium tert-butoxide, 99%, pure
Sodium tert-butoxide, 99%, pure
Sodium tert-butoxide, 99%, pure
Acros Organics

Sodium tert-butoxide, 99%, pure

CAS: 865-48-5 | C4H9NaO | 96.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 252555000
also known as 25255
HSN 29319090
Price (INR)
27,175.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
27,175.00
Each
Chemical Identifiers
CAS865-48-5
IUPAC Namesodium 2-methylpropan-2-olate
Molecular FormulaC4H9NaO
InChI KeyMFRIHAYPQRLWNB-UHFFFAOYSA-N
SMILES[Na+].CC(C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Powder
NaOH + Na2CO3 (as NaOH)=<1.5 %
Titration with HCl98.5 to 101.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only