Nunc™ Disposable Loops and Needles
Actual product may vary
Nunc™ Disposable Loops and Needles
Nunc™ Disposable Loops and Needles
Nunc™ Disposable Loops and Needles
Nunc™ Disposable Loops and Needles
Nunc
Nunc™ Disposable Loops and Needles
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolumeQuantity
2532871 μLCarton of 1000
25158610 μLCarton of 1000
2544101 μLCarton of 600
25443710 μLCarton of 600
Catalog number 253287
HSN 39269099
Price (INR)
47,989.00
Case
Volume:
1 μL
Quantity:
Carton of 1000
Price (INR)
47,989.00
Case
Easily identify color-coded Thermo Scientific™ Nunc™ Disposable Loops and Needles, precision molded from nontoxic (USP Class VI) polystyrene material to provide a smooth loop surface. Thermo Scientific Nunc products are high quality plastics for use in biotechnology, pharmaceutical and research laboratories, as well as in the production of vaccines and diagnostic kits.
Features:
  • Radiation sterilized—no need for flame sterilization
  • Hydrophilic treatment during manufacturing process
  • Liquid adherence properties that prevent accidental discharge of droplets
  • Loops available in 10 and 1 μL sizes
  • Meet semi-quantitative standard for handling samples
  • Calibration documents available for any batch upon request
Specifications
DisposableSingle-use
MaterialPolystyrene
SterilitySterile
Volume1 μL
ColorClear
Product TypeInoculating Loop (Clear)
QuantityCarton of 1000
Unit SizeCase