Product Image
Applied Biosystems™

SPR, Wave Nickel Plated 1.80 OD

This is a Procise Protein Sequencer Spare Part.
Have Questions?
Catalog NumberQuantity
25711 Unit
Catalog number 2571
Price (INR)
-
Quantity:
1 Unit

This is a Procise Protein Sequencer Spare Part.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Equipment)Procise Protein Sequencer
Quantity1 Unit
Shipping ConditionRoom Temperature
Product TypeProtein Sequencer SPR
Unit SizeEach
Contents & Storage
Store at room temperature.