Acetonitrile, 99.5%, for analysis
Acetonitrile, 99.5%, for analysis
Acetonitrile, 99.5%, for analysis
Acros Organics

Acetonitrile, 99.5%, for analysis

CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 258560025
also known as 25856
HSN 29269000
Price (INR)
23,316.00
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
23,316.00
Each
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Calcium (Ca)=<1 ppm
Magnesium (Mg)=<0.5 ppm
Water=<0.1 % (Coulometric)
Lead (Pb)=<2 ppm
Iron (Fe)=<2 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only