Bacto™ TC Lactalbumin Hydrolysate
Gibco™

Bacto™ TC Lactalbumin Hydrolysate

Bacto TC Lactalbumin Hydrolysate, an animal origin peptone, is the enzymatically hydrolyzed protein portion of milk whey. This supplement isRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
25996110 kg
259962500 g
Catalog number 259961
Price (INR)
239,157.00
Each
Quantity:
10 kg
Price (INR)
239,157.00
Each
Bacto TC Lactalbumin Hydrolysate, an animal origin peptone, is the enzymatically hydrolyzed protein portion of milk whey. This supplement is a mixture of peptides, amino acids, and carbohydrates, both simple and complex. Customers have successfully used Bacto TC Lactalbumin Hydrolysate to support the growth of many mammalian, bacterial, and insect cell types, and it has been used to produce both recombinant proteins and animal health vaccines. Customers have also used this supplement as an amino acid supplement for mammalian cell culture media.

Specifications
Culture TypeMammalian Cell Culture
For Use With (Application)Mammalian, bacterial, insect cell types, recombinant proteins and animal health vaccines
Product LineBacto
Quantity10 kg
SourceAnimal Origin
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.