Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates
Actual product may vary
Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates
Improve efficiency in mixing, washing and recovery of samples with Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates.
Have Questions?
Catalog NumberSterilityLidNo. per Pack
262146Non-sterileWithout Lid135
Catalog number 262146
Price (INR)
33,448.00
CS
Sterility:
Non-sterile
Lid:
Without Lid
No. per Pack:
135
Price (INR)
33,448.00
CS
Improve efficiency in mixing, washing and recovery of samples with Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates. The clear and U shaped wells are ideal for solution-based assays (e.g. agglutination assays) and common serial dilutions, allowing clean and easy pipetting. Available in a variety of options for surfaces, sterility, closures, and packaging sizes.
  • Clear polystyrene has excellent optical properties ideal for optical measurements
  • Thermo Scientific™ Nunclon™ Delta is a hydrophilic surface treatment whereas the non-treated polystyrene is more hydrophobic
  • Customized bar coding is available
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceAmerican National Standards (ANSI) (ANSI/SBS 1-2004)
FormatStandard
LidWithout Lid
Array8 x 12
Packaging135 Pk.
Product LineNunc
ColorClear
Product TypeMicroplate
MaterialPolystyrene
No. of Wells96
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeRound Bottom (U-well)
No. per Case135
No. per Pack135
Unit SizeCS