trans-(+)-Chrysanthemic acid, 99+%
trans-(+)-Chrysanthemic acid, 99+%
trans-(+)-Chrysanthemic acid, 99+%
trans-(+)-Chrysanthemic acid, 99+%
Acros Organics

trans-(+)-Chrysanthemic acid, 99+%

CAS: 4638-92-0 | C10H16O2 | 168.23 g/mol
Have Questions?
Catalog NumberQuantity
263635000
also known as 26363
500 mg
Catalog number 263635000
also known as 26363
HSN 29162000
Price (INR)
36,738.00
Each
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (INR)
36,738.00
Each
Add to cart
Chemical Identifiers
CAS4638-92-0
IUPAC Name(1S,3R)-2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropane-1-carboxylic acid
Molecular FormulaC10H16O2
InChI KeyXLOPRKKSAJMMEW-HTQZYQBOSA-N
SMILESCC(C)=C[C@@H]1[C@H](C(O)=O)C1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4760 to 1.4790 (20°C, 589 nm)
GC>=99.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only