Search
| Catalog Number | Color | Sterility | Well Shape | Fit for Purpose |
|---|---|---|---|---|
| 264574 | Natural | Sterile | Square, Round Bottom (U-well) | Certified Quality |
| 264573 | Natural | Non-sterile | Square, Round Bottom (U-well) | Certified Quality |
| 264575 | White | Non-sterile | Square, Conical Bottom (V-well) | Certified Quality |
| 264576 | Black | Non-sterile | Square, Round Bottom (U-well) | Certified Quality |
| 264579 | Blue | Non-sterile | Square, Round Bottom (U-well) | Certified Quality |
| 4309 | Natural | Sterile | Square, Conical Bottom (V-well) | Certified Quality |
| 4308 | Black | Non-sterile, Non-sterile | Square, Conical Bottom (V-well) | Certified Quality |
| 4312 | Natural | Non-sterile | Square-Rounded | Certified Quality |
Minimize costs when storing compounds for small volume applications. Thermo Scientific™ 384-Well Conical Bottom Polypropylene Plates feature a lower binding capacity so proteins or DNA will not bind, allowing for complete sample recovery. Withstands temperatures from -80° to +121°C. Offered in a variety of colors for quick identification of your storage plates.