3-Fluoro-1,2-dihydroxybenzene, 98+%, Thermo Scientific Chemicals
3-Fluoro-1,2-dihydroxybenzene, 98+%, Thermo Scientific Chemicals
3-Fluoro-1,2-dihydroxybenzene, 98+%, Thermo Scientific Chemicals
Acros Organics

3-Fluoro-1,2-dihydroxybenzene, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number 264790100
also known as 26479
HSN 29062990
Price (INR)
27,317.00
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
27,317.00
Each
Chemical Identifiers
CAS363-52-0
IUPAC Name3-fluorobenzene-1,2-diol
Molecular FormulaC6H5FO2
InChI KeyDXOSJQLIRGXWCF-UHFFFAOYSA-N
SMILESOC1=CC=CC(F)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.0 %
Appearance (Color)Light beige to grey-brown
Infrared spectrumConforms
Melting point69°C to 73°C
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only