trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride, 99+%
trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride, 99+%
trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride, 99+%
Acros Organics

trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride, 99+%

CAS: 28250-45-5 | C7H16NO | 130.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Packaging:
Glass bottle
Catalog number 265870010
also known as 26587
HSN 29213010
Price (INR)
45,639.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
45,639.00
Each
Chemical Identifiers
CAS28250-45-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration Argentometric>=99.0 %
Appearance (Form)Crystalline powder
Melting point150°C to 155°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only