96 Well Plate, Non-Treated Surface, Pack of 25
Thermo Scientific™
96 Well Plate, Non-Treated Surface, Pack of 25
These microplates are optimized for robotics, automated readers and liquid handling systems. Ideal for antibiotic screens, cell-based assays and screening compounds.
Have Questions?
Catalog NumberQuantity
266120100/Cs.
Catalog number 266120
Price (INR)
45,000.00
Case of 100
Quantity:
100/Cs.
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Description96F well plate, Non-Treated, clear, with lid, Sterile (Pack of 25, Case of 100)
LidWith Lid
No. per Case100
No. per Pack25
Packaging25/Pk., 100/Cs.
Product TypeMicroplate
Quantity100/Cs.
Array8 x 12
BarcodeCustom Barcoding Available
ColorClear
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilitySterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 100