5-Sulfosalicylic acid dihydrate, 99+%
5-Sulfosalicylic acid dihydrate, 99+%
5-Sulfosalicylic acid dihydrate, 99+%
Acros Organics

5-Sulfosalicylic acid dihydrate, 99+%

CAS: 5965-83-3 | C9H14O6S | 250.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 268792500
HSN 29182990
Price (INR)
7,989.00
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
7,989.00
Each
Chemical Identifiers
CAS5965-83-3
IUPAC Name2-hydroxy-5-sulfobenzoic acid dihydrate
Molecular FormulaC7H10O8S
InChI KeyBHDKTFQBRFWJKR-UHFFFAOYSA-N
SMILESO.O.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH99.0 to 101.0 %
Heavy metals (as Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only