4-Pentenoic acid, 99%
4-Pentenoic acid, 99%
4-Pentenoic acid, 99%
4-Pentenoic acid, 99%
Acros Organics

4-Pentenoic acid, 99%

CAS: 591-80-0 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 269021000
also known as 26902
HSN 29159090
Price (INR)
31,436.00
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
31,436.00
Each
Add to cart
Chemical Identifiers
CAS591-80-0
IUPAC Namepent-4-enoic acid
Molecular FormulaC5H8O2
InChI KeyHVAMZGADVCBITI-UHFFFAOYSA-N
SMILESOC(=O)CCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Specific gravity(25°C) 0.971 to 0.979
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only