1-Amino-3,3-diethoxypropane, 97%
1-Amino-3,3-diethoxypropane, 97%
1-Amino-3,3-diethoxypropane, 97%
1-Amino-3,3-diethoxypropane, 97%
Acros Organics

1-Amino-3,3-diethoxypropane, 97%

CAS: 41365-75-7 | C7H17NO2 | 147.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 269851000
also known as 26985
HSN 29221990
Price (INR)
64,789.00
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
64,789.00
Each
Add to cart
Chemical Identifiers
CAS41365-75-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive index1.4233 to 1.4253 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only