2'-Fluoro-Bz-A Phosphoramidite, standard grade, serum vial bottle
Thermo Scientific™

2'-Fluoro-Bz-A Phosphoramidite, standard grade, serum vial bottle

2'-Fluoro-Bz-A CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with 2'-Fluoro protection, for use in oligonucleotideRead more
Have Questions?
Catalog NumberQuantity
27-1601-021 g
Catalog number 27-1601-02
Price (INR)
0.01
Each
Quantity:
1 g
Price (INR)
0.01
Each

2'-Fluoro-Bz-A CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with 2'-Fluoro protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, the highly electronegative fluorine (2'-Fluoro) modification adopts a more thermodynamically stable A-form helix on hybridization to its target. Showing improved affinity and some nuclease resistance (compared to 2'-OH), the 2'-F modification has been proven useful in antisense, siRNA, and aptamer applications. The 'standard grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 20-mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N6-benzoyl-2'-fluoroadenosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C47H51FN7O7P
• Base—A
• Base Protection—Bz
• Formula weight—875.92 g/mol
• HPLC purity—>98%
31P NMR purity—≥98%

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AKTA Synthesizers, Applied Biosystems Synthesizers
FormPowder
GradeStandard
Nucleic Acid TypeRNA
Nucleoside TypeAdenosine (A)
Product Type2in-Fluoro-Bz-A CEP
Purity98%
Quantity1 g
Reagent Features2in-Fluoro modified
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).