TheraPure™ 2'-Fluoro-U Phosphoramidite, serum vial bottle
TheraPure™ 2'-Fluoro-U Phosphoramidite, serum vial bottle
Thermo Scientific™

TheraPure™ 2'-Fluoro-U Phosphoramidite, serum vial bottle

2'-Fluoro-U CEP is a phosphoramidite containing 5' DMT and the nucleoside uridine (U) with 2'-Fluoro protection, for use in oligonucleotideRead more
Have Questions?
Catalog NumberQuantity
27-1602-021 g
Catalog number 27-1602-02
Price (INR)
0.01
Each
Add to cart
Quantity:
1 g
Price (INR)
0.01
Each
Add to cart

2'-Fluoro-U CEP is a phosphoramidite containing 5' DMT and the nucleoside uridine (U) with 2'-Fluoro protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, the highly electronegative fluorine (2'-Fluoro) modification adopts a more thermodynamically stable A-form helix on hybridization to its target. Showing improved affinity and some nuclease resistance (compared to 2'-OH), the 2'-F modification has been proven useful in antisense, siRNA, and aptamer applications. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 20-mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-2'-fluorouridine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C39H46FN4O8P
• Base—A
• Formula weight—748.8 g/mol
• CAS number—146954-75-8
• HPLC purity—>98%
31P NMR purity—≥98%

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AKTA Synthesizers, Applied Biosystems Synthesizers
FormPowder
GradeTheraPure™
Nucleic Acid TypeRNA
Nucleoside TypeUridine (U)
Product Type2in-Fluoro-U CEP
Purity98%
Quantity1 g
Reagent Features2in-Fluoro modified
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Product LineTheraPure
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).