Bz-dC Phosphoramidite, standard grade, 20-400 finish bottle
Thermo Scientific™

Bz-dC Phosphoramidite, standard grade, 20-400 finish bottle

Bz-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Benzoyl (Bz) base protection, for useRead more
Have Questions?
Catalog NumberQuantity
27-1732-112 g
Catalog number 27-1732-11
Price (INR)
0.01
Each
Quantity:
2 g
Price (INR)
0.01
Each

Bz-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Benzoyl (Bz) base protection, for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 30 mL Amber bottle with a 20-400 Finish neck style, which is compatible with most Expedite Systems of oligonucleotide synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N4-benzoyl-2'-deoxycytidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C46H52N5O8P
• Base—dC
• Base Protection—Bz
• Formula weight—833.93 g/mol
• CAS number—102212-98-6
• HPLC Purity—≥98.0%
31P NMR Purity—≥98.0%

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Expedite Synthesizers
FormPowder
GradeStandard
Nucleic Acid TypeDNA
Nucleoside TypeDeoxycytidine (dC)
Product TypeBz-dC CEP
Purity98%
Quantity2 g
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).