HEG Phosphoramidite, standard grade, serum vial bottle
Thermo Scientific™

HEG Phosphoramidite, standard grade, serum vial bottle

HEG CEP (Spacer 18) is a hexaethyleneglycol-based phosphoramidite for use in oligonucleotide synthesis. This unique property creates a hydrophilic spacer,Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
27-1786-040.25 g
27-1786-021 g
Catalog number 27-1786-04
Price (INR)
0.01
Each
Quantity:
0.25 g
Price (INR)
0.01
Each

HEG CEP (Spacer 18) is a hexaethyleneglycol-based phosphoramidite for use in oligonucleotide synthesis. This unique property creates a hydrophilic spacer, which can be useful for building flexibility within an oligo sequence. It has proven useful for probes requiring minimal steric hindrance (usually when conjugated to an antibody or enzyme), multiple binding sites, and polymerase activity at the 3' terminus to be eliminated. This product is supplied in a 20 mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers of oligonucleotide synthesizers.

Features:
• Full chemical name—18-O-(4,4'-Dimethoxytrityl)-Hexaethyleneglycol-1-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C42H61N2O10P
• Formula weight—784.9 g/mol
• HPLC Purity—≥95.0%; Bis impurity ≤0.3%; Penta impurity ≤1.6%; Hepta impurity ≤0.9%
31P NMR Purity—≥98%; Sum of non-primary peaks in 140-152 ppm range: ≤0.5%

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AKTA Synthesizers, Applied Biosystems Synthesizers
FormOil
GradeStandard
Nucleic Acid TypeNone (Modifier)
Nucleoside TypeNone (Modifier)
Product TypeHEG CEP
Purity95%
Quantity0.25 g
Reagent TypePhosphoramidite
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).