2'-OMe-iBu-G Phosphoramidite, standard grade, serum vial bottle
Thermo Scientific™

2'-OMe-iBu-G Phosphoramidite, standard grade, serum vial bottle

2'-OMe-iBu-G CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with 2'-O-methyl protection, for use in oligonucleotideRead more
Have Questions?
Catalog NumberGrade
27-1846-02Standard
Catalog number 27-1846-02
Price (INR)
0.01
Each
Grade:
Standard
Price (INR)
0.01
Each
2'-OMe-iBu-G CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with 2'-O-methyl protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methyl (OMe) oligo modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The 'standard grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 20-mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methyl-guanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C45H56N7O9P
• Base—G
• Base Protection—iBu
• Formula weight—869.9 g/mol
• CAS number—150780-67-9
• HPLC purity—≥98.0%; Sum of non-primary peaks 140-152ppm: ≤0.5%
31P NMR purity—≥98.0%

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AKTA Synthesizers, Applied Biosystems Synthesizers
FormatPowder
GradeStandard
Nucleic Acid TypeRNA
Nucleoside TypeGuanosine (G)
Product Type2'-OMe-iBu-G CEP
Purity98%
Reagent Features2'-OMe modified
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).