Product Image
Thermo Scientific™

TheraPure™ Ac-dC Phosphoramidite, 28-400 finish bottle

Ac-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Acetyl (Ac) base protection, for useRead more
Have Questions?
Catalog NumberQuantity
27-2027-1410 g
Catalog number 27-2027-14
Price (INR)
0.01
Each
Quantity:
10 g
Price (INR)
0.01
Each

Ac-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Acetyl (Ac) base protection, for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. By using Ac-dC, Bz-dA, and DMF-dG monomers, fast cleavage and deprotection can be effected by a 1:1 mixture of aqueous ammonium hydroxide and aqueous methylamine (or AMA) in 10 min. This product is supplied in a 250 mL Amber bottle with a 28-400 Finish neck style, which is compatible with most MerMade Systems of oligonucleotide synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N4-Acetyl-2'-deoxycytidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)-phosphoramidite
• Molecular formula—C41H50N5O8P
• Base—dC
• Base Protection—Ac
• Formula weight—771.8 g/mol
• CAS number—154110-40-4
• HPLC Purity—≥99%; M-11 Impurity ≤0.01%; No single impurity >0.5%
31P NMR Purity—≥99%; Sum of non-primary peaks at 140-152 ppm: ≤0.3%

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)MerMade Synthesizers
FormPowder
GradeTheraPure™
Nucleic Acid TypeDNA
Nucleoside TypeDeoxycytidine (dC)
Product TypeAc-dC CEP
Purity99%
Quantity10 g
Reagent FeaturesFast deprotection
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Product LineTheraPure
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).