TheraPure™ 2'-OMe-C Phosphoramidite, serum vial bottle
Thermo Scientific™

TheraPure™ 2'-OMe-C Phosphoramidite, serum vial bottle

2'-OMe-C CEP is a phosphoramidite containing 5' DMT and the nucleoside cytidine (C) with 2'-O-methyl protection, for use in oligonucleotideRead more
Have Questions?
Catalog NumberQuantity
27-2043-041 g
Catalog number 27-2043-04
Price (INR)
0.01
Each
Quantity:
1 g
Price (INR)
0.01
Each

2'-OMe-C CEP is a phosphoramidite containing 5' DMT and the nucleoside cytidine (C) with 2'-O-methyl protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methyl (OMe) oligo modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 20-mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N4-acetyl-2'-O-methyl-cytidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C42H52N5O9P
• Base—C
• Base Protection—Ac
• Formula weight—801.86 g/mol
• CAS number—199593-09-4
• HPLC purity—≥99%; M-11 Impurity: ≤0.01%; Single critical impurity ≤0.3%
31P NMR purity—>99%; Sum of non-primary peaks at 140-152 ppm ≤0.5%

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AKTA Synthesizers, Applied Biosystems Synthesizers
FormPowder
GradeTheraPure™
Nucleic Acid TypeRNA
Nucleoside TypeCytidine (C)
Product Type2in-OMe-C CEP
Purity99%
Quantity1 g
Reagent Features2in-OMe modified
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Product LineTheraPure
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).