1-Pentanesulfonic acid, sodium salt hydrate, 99%
1-Pentanesulfonic acid, sodium salt hydrate, 99%
1-Pentanesulfonic acid, sodium salt hydrate, 99%
1-Pentanesulfonic acid, sodium salt hydrate, 99%
1-Pentanesulfonic acid, sodium salt hydrate, 99%
1-Pentanesulfonic acid, sodium salt hydrate, 99%
Acros Organics

1-Pentanesulfonic acid, sodium salt hydrate, 99%

CAS: 1266615-65-9 | C5H11NaO3S | 174.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 270371000
also known as 27037
HSN 29041090
Price (INR)
45,070.00
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
45,070.00
Each
Add to cart
Chemical Identifiers
CAS1266615-65-9
IUPAC Namesodium pentane-1-sulfonate
Molecular FormulaC5H11NaO3S
InChI KeyROBLTDOHDSGGDT-UHFFFAOYSA-M
SMILES[Na+].CCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(10 % in water) Clear colorless
Appearance (Form)Crystalline powder
Appearance (Color)White
Infrared spectrumConforms
Water=<9.6 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only