2,3,4,6-Tetra-O-acetyl-alpha-D-glucopyranosyl bromide, 98.5%
2,3,4,6-Tetra-O-acetyl-alpha-D-glucopyranosyl bromide, 98.5%
2,3,4,6-Tetra-O-acetyl-alpha-D-glucopyranosyl bromide, 98.5%
2,3,4,6-Tetra-O-acetyl-alpha-D-glucopyranosyl bromide, 98.5%
Acros Organics

2,3,4,6-Tetra-O-acetyl-alpha-D-glucopyranosyl bromide, 98.5%

CAS: 572-09-8 | C14H19BrO9 | 411.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 270930500
also known as 27093
HSN 29400000
Price (INR)
46,917.00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
46,917.00
Each
Chemical Identifiers
CAS572-09-8
IUPAC Name[(2R,3R,4S,5R,6R)-3,4,5-tris(acetyloxy)-6-bromooxan-2-yl]methyl acetate
Molecular FormulaC14H19BrO9
InChI KeyCYAYKKUWALRRPA-HAGGZJDPNA-N
SMILESCC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration Argentometric>=98.0 %
Specific optical rotation+188° to +202° (20°C, 589 nm) (c=2, CHCl3)
Appearance (Form)Powder
Appearance (Color)White to beige to light grey
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only