Metaldehyde, 99%
Metaldehyde, 99%
Metaldehyde, 99%
Acros Organics

Metaldehyde, 99%

CAS: 108-62-3 | C8H16O4 | 176.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number 271862500
HSN 29329900
Price (INR)
13,658.00
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
13,658.00
Each
Chemical Identifiers
CAS108-62-3
IUPAC Name2,4,6,8-tetramethyl-1,3,5,7-tetraoxocane
Molecular FormulaC8H16O4
InChI KeyGKKDCARASOJPNG-UHFFFAOYSA-N
SMILESCC1OC(C)OC(C)OC(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only