1,2,3,4-Tetrahydro-2-naphthol, 97%
1,2,3,4-Tetrahydro-2-naphthol, 97%
1,2,3,4-Tetrahydro-2-naphthol, 97%
Acros Organics

1,2,3,4-Tetrahydro-2-naphthol, 97%

CAS: 530-91-6 | C10H12O | 148.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 271880050
also known as 27188
HSN 29089990
Price (INR)
63,818.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
63,818.00
Each
Chemical Identifiers
CAS530-91-6
IUPAC Name1,2,3,4-tetrahydronaphthalen-2-ol
Molecular FormulaC10H12O
InChI KeyJWQYZECMEPOAPF-UHFFFAOYNA-N
SMILESOC1CCC2=CC=CC=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Viscous liquid
Appearance (Color)Clear light yellow to light brown
Refractive index1.5630 to 1.5670 (20°C, 589 nm)
AppearanceMay darken on storage
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only