BOC-L-Phenylalanine, 99+%
BOC-L-Phenylalanine, 99+%
BOC-L-Phenylalanine, 99+%
Acros Organics

BOC-L-Phenylalanine, 99+%

CAS: 13734-34-4 | C14H19NO4 | 265.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass Bottle
Catalog number 275640250
HSN 29224990
Price (INR)
9,587.00
Each
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS13734-34-4
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoic acid
Molecular FormulaC14H19NO4
InChI KeyZYJPUMXJBDHSIF-NSHDSACASA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point85°C to 88°C
Specific optical rotation+24° to +26° (20°C, 589 nm) (c=1, ethanol)
Titration with NaOH>=99.0 %
Water=<0.5 % (K.F.)
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only