2-Amino-1-methylbenzimidazole, 98+%
2-Amino-1-methylbenzimidazole, 98+%
2-Amino-1-methylbenzimidazole, 98+%
Acros Organics

2-Amino-1-methylbenzimidazole, 98+%

CAS: 1622-57-7 | C8H9N3 | 147.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
276300025
also known as 27630
2.5 gGlass bottle
Catalog number 276300025
also known as 27630
HSN 29339900
Price (INR)
16,546.00
Each
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
16,546.00
Each
Chemical Identifiers
CAS1622-57-7
IUPAC Name1-methyl-1H-1,3-benzodiazol-2-amine
Molecular FormulaC8H9N3
InChI KeyXDFZKQJLNGNJAN-UHFFFAOYSA-N
SMILESCN1C(N)=NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.0 %
Appearance (Form)Crystalline powder
Melting point203°C to 206°C
Appearance (Color)White to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only