SoftFit-L™ Pipette Tips, Filtered, Low-Retention, in Racks with Lift-off Lids

SoftFit-L™ Pipette Tips, Filtered, Low-Retention, in Racks with Lift-off Lids

Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ Filtered Pipette Tips in Racks in Lift-off Lids. For use with Rainin™ LTS pipettes.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
2769-05PK200 μL
2779-05PK1000 μL
2749-05PK20 μL
Catalog number 2769-05PK
Price (INR)
-
Volume (Metric):
200 μL
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
  • Low-retention tips maximize sample delivery
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, and Endotoxin Free
Description200L, Filtered, Sterile, Rack
Length (English)1.97 in.
Length (Metric)5.00cm
Model200L
ColorNatural
Color CodeGreen
FilteredFiltered
For Use With (Equipment)Rainin LTS LiteTouch
FormatRack
Product LineART SoftFit L
SterilitySterile
Tip StyleUniversal, Low Retention
Volume (Metric)200 μL
Unit SizePack of 960