SoftFit-L™ Filtered Pipette Tips in Hinged Racks
SoftFit-L™ Filtered Pipette Tips in Hinged Racks
SoftFit-L™ Filtered Pipette Tips in Hinged Racks
SoftFit-L™ Filtered Pipette Tips in Hinged Racks
SoftFit-L™ Filtered Pipette Tips in Hinged Racks

SoftFit-L™ Filtered Pipette Tips in Hinged Racks

Pipette samples with the accuracy and precision you expect from your pipette, every time with these pipette tips. For use with Rainin™ LTS pipettes.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
2779-HRPK
also known as 2779-HR
1000 μL
2749-HRPK
also known as 2749-HR
20 μL
2739-HRPK
also known as 2739-HR
300 μL
2769-HRPK
also known as 2769-HR
200 μL
2789-HRPK
also known as 2789-HR
1200 μL
Catalog number 2779-HRPK
also known as 2779-HR
Price (INR)
-
Volume (Metric):
1000 μL
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
  • Hinged rack provides easy one-hand access to tips
  • Color-coded inserts allow quick identification of tip volume
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, and Endotoxin Free
Description1000 μL, Filtered, Sterile, Hinged Rack
Length (English)3.39 in.
Length (Metric)8.61cm
Model1000L
ColorNatural
Color CodeBlue
FilteredFiltered
Fit for PurposeSterile & PCR Ready
For Use With (Equipment)Rainin LTS LiteTouch
FormatHinged Rack
MaterialPolypropylene
Product LineART SoftFit L
SterilitySterile
Tip StyleUniversal
Volume (Metric)1000 μL
Unit SizePack of 768