Search
Search
Perform pipetting accurately and consistently with these barrier extended length pipette tips, which seal when exposed to potential contaminants.
| Catalog Number | Volume (Metric) | Model | Tip Style | Fit for Purpose | Unit Size |
|---|---|---|---|---|---|
| 2780PK also known as 2780 | 50 μL | ART 50U | Ergonomic | Sterile and PCR Ready | Pack of 960 |
| 2380 | 1000 μL | ART 1000 ERGO-F | Ergonomic | Sterile and PCR Ready | Each |
| 2279PK also known as 2279 | 1000 μL | ART 1000 | Ergonomic | Sterile and PCR Ready | Pack of 800 |
| 2140 | 10 μL | ART 10 REACH | Extended Length | Sterile and PCR Ready | Case of 4800 |
| 2140PK also known as 2140 | 10 μL | ART 10 REACH | Extended Length | Sterile and PCR Ready | Pack of 960 |
| 2149PK also known as 2149 | 0.5 to 20 μL | ART 20E | Micro Point | Sterile and PCR Ready | Pack of 960 |
Features:
Designed to Aid in Pipetting for a Variety of Applications:
Extended length: to prevent contamination by keeping the pipette shaft from entering the sample vessel
Gel loading: for loading acrylamide or agarose gels
Wide orifice genomic: for genomic DNA, fragile cell lines and other viscous materials
Solvent safe: for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson Pipetman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes