1,4-Dibromo-2,5-dimethylbenzene, 98+%
1,4-Dibromo-2,5-dimethylbenzene, 98+%
1,4-Dibromo-2,5-dimethylbenzene, 98+%
Acros Organics

1,4-Dibromo-2,5-dimethylbenzene, 98+%

CAS: 1074-24-4 | C8H8Br2 | 263.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 279010500
also known as 27901
HSN 29049090
Price (INR)
12,238.00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
12,238.00
Each
Chemical Identifiers
CAS1074-24-4
IUPAC Name1,4-dibromo-2,5-dimethylbenzene
Molecular FormulaC8H8Br2
InChI KeyQENIALCDPFDFHX-UHFFFAOYSA-N
SMILESCC1=CC(Br)=C(C)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to light orange
Melting point72°C to 76°C
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only