Gallic acid ethyl ester, 99%
Gallic acid ethyl ester, 99%
Gallic acid ethyl ester, 99%
Acros Organics

Gallic acid ethyl ester, 99%

CAS: 831-61-8 | C9H10O5 | 198.174 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 279931000
also known as 27993
HSN 29182990
Price (INR)
10,084.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
10,084.00
Each
Chemical Identifiers
CAS831-61-8
IUPAC Nameethyl 3,4,5-trihydroxybenzoate
Molecular FormulaC9H10O5
InChI KeyVFPFQHQNJCMNBZ-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC(O)=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=98.5 %
Melting point151°C to 154°C
Appearance (Color)White to light beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only