Hypersil™ BDS C18 Columns
Hypersil™ BDS C18 Columns
Actual product may vary

Hypersil™ BDS C18 Columns

Use as a robust, general-purpose column for a wide range of applications in QC/QA labs.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)Particle SizeColumn Format
28103-01300110 mm3 mm3 μmGuard Cartridge, 4 Pk.
Catalog number 28103-013001
HSN 90279090
Price (INR)
59,738.00
Pack
Add to cart
Length (Metric):
10 mm
Diameter (Metric):
3 mm
Particle Size:
3 μm
Column Format:
Guard Cartridge, 4 Pk.
Price (INR)
59,738.00
Pack
Add to cart

For a robust, general-purpose column to use in a wide range of applications, choose Thermo Scientific™ Hypersil™ BDS C18 HPLC Columns. These economical columns feature reduced silanol activity and provide exceptional reliability for QA/QC labs. Built on the renowned Hypersil silica backbone, the base-deactivated silica is one of the most popular packing materials available.

For more challenging separations that require improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ columns as an alternative.

  • Original Hypersil columns from the manufacturer
  • Base deactivated with minimal residual silanol activity
  • Exceptional reproducibility
  • Economical, general purpose columns

Ordering Information:

Other column dimensions are available. Please call Customer Service for more information.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatGuard Cartridge, 4 Pk.
Diameter (Metric)3 mm
Length (Metric)10 mm
Max. Pressure5800 psi (400 bar)
Particle Size3 μm
pH2 to 9
Pore Size130 Å
Surface Area170 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LineHypersil BDS
Packing MaterialSpherical, Fully Porous, Base-Deactivated Silica
Carbon Load11%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit SizePack