Hypersil™ BDS C18 Column, 5um
Actual product may vary

Hypersil™ BDS C18 Column, 5um

For a robust, general-purpose column to use in a wide range of applications, choose Thermo Scientific™ Hypersil™ BDS C18 HPLC Columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
28105-3040304 mm
28105-3046304.6 mm
Catalog number 28105-304030
HSN 90279090
Price (INR)
83,068.00
Each
Diameter (Metric):
4 mm
Price (INR)
83,068.00
Each
  • Base-deactivated with minimal residual silanol activity
  • Exceptional reproducibility
  • Economical, general purpose columns
Specifications
Column FormatAnalytical Column
Diameter (Metric)4 mm
Length (Metric)300 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size130 Å
Surface Area170 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Packing MaterialSpherical, Fully Porous, Base-Deactivated Silica
Carbon Load11%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit SizeEach