POROS™ CaptureSelect™ IgM-XL Affinity Matrix
Thermo Scientific™

POROS™ CaptureSelect™ IgM-XL Affinity Matrix

Thermo Scientific™ POROS™ CaptureSelect™ IgM-XL Affinity Matrix is intended for the purification of human IgM, with no binding to other immunoglobulins.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
28128920055 mL
281289201010 mL
281289205050 mL
2812892500500 mL
Catalog number 2812892005
Price (INR)
-
Quantity:
5 mL

POROS CaptureSelect IgM-XL Affinity Matrix is intended for the purification of human IgM, with no binding to other immunoglobulins.

Features of this affinity matrix include:

  • IgM purification from plasma and serum
  • Purification of monoclonal IgM from cell cultures
  • Efficient purification of IgM with high yields and retained activity
  • Excellent scalability
  • Non-animal derived
  • High flow rates
  • Cross-reactivity towards mouse and rat IgM

POROS CaptureSelect IgM-XL Affinity Matrix binds to the Fc part of IgM. The high selectivity and yields obtained using this affinity matrix enable a robust, fast, and efficient purification process with excellent purity obtained in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (animal origin-free).

Main characteristics

  • Matrix: cross-linked poly(styrene-divinylbenzene)
  • Average particle size: 50 μm
  • Ligand: POROS CaptureSelect IgM-XL affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >5 g/L resin (determined with polyclonal human IgM from serum)
  • Elution conditions: acidic elution with 0.1 M glycine, pH 3.0
  • Mechanical resistance: 100 bar (1450 psi, 10 MPa)
  • Formulation buffer: 20% (v/v) ethanol
Specifications
Binding Capacity>5 g/L
Ligand TypeCaptureSelect IgM-XL Affinity Ligand
MatrixCross-linked poly(styrene-divinylbenzene) POROS beads
Max. Operating Pressure100 bar
Quantity5 mL
Shipping ConditionWet Ice
Stationary Phase20% Ethanol
Capacity (Metric)>5 g/L
FormResin Slurry
Particle Size50 μm
Product LinePOROS CaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C in a light-protected environment. Avoid direct exposure to sunlight. Do not freeze