Sodium Dodecyl Sulfate (SDS), Lauryl
Sodium Dodecyl Sulfate (SDS), Lauryl
Sodium Dodecyl Sulfate (SDS), Lauryl
Thermo Scientific™

Sodium Dodecyl Sulfate (SDS), Lauryl

Perform routine protein electrophoresis and cell lysis methods using this popular, standard-grade SDS detergent.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
283651 kg
28364100 g
Catalog number 28365
Price (INR)
35,693.00
Online offer
Ends: 31-Dec-2026
52,878.00
Save 17,185.00 (32%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (INR)
35,693.00
Online offer
Ends: 31-Dec-2026
52,878.00
Save 17,185.00 (32%)
Each

Thermo Scientific Sodium Dodecyl Sulfate (Lauryl) is standard-grade SDS detergent for use in protein polyacrylamide gel electrophoresis (PAGE).

Features of Sodium Dodecyl Sulfate (Lauryl):

  • Popular anionic detergent for a variety of protein methods
  • Especially useful for denaturing polyacrylamide gel electrophoresis (SDS-PAGE)
  • Common component of cell lysis buffers

This lauryl-grade sodium dodecyl sulfate (SDS) is a popular anionic detergent for routine protein electrophoresis and cell lysis methods. The formulation is a mixture of several different alkyl sulfate chain lengths (C10 to C18).

Properties of SDS (values for pure C12):

  • Molecular Weight: 288.5 g
  • Detergent Class: Ionic (anionic)
  • Aggregation Number: 62
  • Micelle Molecular Weight: 18,000 g
  • Critical Micelle Concentration (CMC): 6 to 8 mM (0.1728 to 0.2304%, w/v)
  • Cloud Point: >100°C
  • Dialyzable: No

Specifications for Sodium Dodecyl Sulfate:

  • Visual: White powder, free of foreign material.
  • Solubility: 10% (aq, w/v) solution must be clear, colorless to slightly yellow.
  • Chain length: C12 >60%; C14 = 20 to 35%; C16 <10%; C10 and C18 <1% each

Related Products

  • Sodium Dodecyl Sulfate (SDS), C12 (Cat. No. 28312)
  • Sodium Dodecyl Sulfate (SDS), 20% Solution (Cat. No. 62202)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorWhite
Quantity1 kg
Reagent TypeDetergent (Pure)
Solubility10% (aq, w/v) solution must be clear, colorless to slightly yellow
FormPowder
Product TypeDetergent
Unit SizeEach
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C.