Dry-blend Buffered Packs
Thermo Scientific™

Dry-blend Buffered Packs

Our quality control ensures that every pack of dry-blend buffers yields consistent solutions without weighing or pH adjustment. Simply empty contents of one foil envelope pack into a beaker, add appropriate solvents, and stir to dissolve.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityChemical Name or Material
2839010 PacksMES Buffered Saline
2837240 PacksPhosphate Buffered Saline (PBS)
2838240 PacksCarbonate-Bicarbonate Buffer Packs
2837640 PacksTris Buffered Saline Packs
2838440 PacksBorate Buffer
2838040 PacksTris-Glycine Buffer
2837910 PacksTris Buffered Saline Packs
2837440 PacksDPBS
Catalog number 28390
Price (INR)
22,914.00
線上優惠
Ends: 31-Dec-2026
32,733.00
Save 9,819.00 (30%)
Each
Quantity:
10 Packs
Chemical Name or Material:
MES Buffered Saline
Request bulk or custom format
Price (INR)
22,914.00
線上優惠
Ends: 31-Dec-2026
32,733.00
Save 9,819.00 (30%)
Each
Our dry-blend buffer packs are easy to use. Simply empty contents of one foil envelope pack into a beaker, add ultrapure water, and stir to dissolve. No need to make and store large volumes of stock solution in advance of daily needs.
Benefits of ready-to-use buffered packs include:
• Convenient—no weighing, no pH adjustment, no need to stock individual components
• Consistent quality—our quality control ensures that every pack yields the same, consistent buffer
• Long shelf life—stocking and storage as dry packs eliminates concerns about long-term stability of stock solutions

BupH™ Phosphate-Buffered Saline Packs are pouches of dry-blend powder that make sodium phosphate buffered saline (PBS) for use in crosslinking and labeling reactions requiring amine-free conditions. Each BupH Dry-Blend pack makes ready-to-use PBS with 0.1 M sodium phosphate, 0.15 M NaCl buffer at pH 7.2 when dissolved in 500 mL of water.

BupH MES-Buffered Saline Packs are pouches of dry-blended powder that make optimized amine-free and carboxyl-free buffer. When dissolved in 500 mL of water, the buffer contains 0.1 M 2-(N-morpholino) ethanesulfonic acid (MES), 0.9% sodium chloride at pH 4.7 for carbodiimide (EDC) crosslinking and conjugation reactions.

BupH Carbonate-Bicarbonate Buffer Packs are pouches of dry-blended powder that are each sufficient to make 500 mL of alkaline (pH 9.4) sodium-carbonate (carb-bicarb) buffer. This buffer is used for coating or binding capture-antibodies and other proteins onto polystyrene microplates for ELISA and other plate assays. When dissolved in 500 mL of water, each pack makes 0.2 M carbonate-bicarbonate at pH 9.4.

BupH Tris-Buffered Saline Packs are pouches of dry-blend powder that make Tris-NaCl buffered saline (TBS) for diluents and wash buffers in ELISA, western blotting, and general immunoassay procedures. Each pack makes ready-to-use TBS with 25 mM Tris, 0.15 M NaCl at pH 7.2 when dissolved in 500 mL of water.

BupH Borate Buffer Packs are pouches of dry-blended powder that are each sufficient to make borate buffer for use in protein modification procedures requiring amine-free conditions. When dissolved in 500 mL of water, each pack makes 50 mM borate at pH 8.5.

BupH Tris-Glycine Transfer Buffer Packs are pouches of dry-blend powder that make 500 mL of standard transfer buffer for wet or semi-dry electrophoretic protein transfer from gel to blotting membrane. Each pack makes 25 mM Tris, 192 mM glycine at pH 8 when dissolved in 400 mL of water and 100 mL methanol.

BupH Modified Dulbecco's PBS Packs are pouches of dry-blend powder that make physiological sodium- and potassium-phosphate buffered saline (D-PBS) for ELISA and western blot diluents and wash buffers. Each pack makes 8 mM sodium phosphate, 2 mM potassium phosphate, 0.14 M NaCl, 10 mM KCl modified Dulbecco's PBS buffer at pH 7.4 when dissolved in 500 mL of water.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialMES Buffered Saline
Recommended StorageStore in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20–25°C.
ConcentrationDry Blend
For Use With (Application)Carboxyl-to-Amine Crosslinking
Physical FormPowder
Product LineBupH
Quantity10 Packs
pH4.7
Unit SizeEach