CaptureSelect™ CH1-XL Magnetic Agarose Beads
Thermo Scientific™

CaptureSelect™ CH1-XL Magnetic Agarose Beads

Thermo Scientific™ CaptureSelect™ CH1-XL Magnetic Agarose Beads are high-capacity, high-throughput magnetic affinity particles for antibody purification using manual or robotic magnetic separators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
28834620011 mL
28834620055 mL
288346205050 mL
Catalog number 2883462001
Price (INR)
52,210.00
Each
Quantity:
1 mL
Price (INR)
52,210.00
Each

CaptureSelect CH1-XL Magnetic Agarose Beads are high-capacity, high-throughput magnetic affinity particles for antibody purification using manual or robotic magnetic separators. The CaptureSelect CH1-XL affinity ligand recognizes the CH1 domain of human IgG, enabling purification of all subclasses (1, 2, 3, and 4) of human IgG and Fab fragments, independent of the light-chain isotype and source material. Due to selectivity for the CH1 domain, no co-purification of free light chains will occur. IgG and Fab fragments can be directly isolated from complex source materials in a single step with high purity and yield.

Features of CaptureSelect CH1-XL Magnetic Agarose Beads include:

  • One-step human IgG purification with high purity, from crude material
  • Compatible with manual and automated workflows (e.g., Thermo Scientific KingFisher instruments)
  • Short operation time
  • Mild elution at pH 4
  • No sample dilution required during isolation, repetitive sample loading feasible
  • No binding to bovine antibodies, allowing use of fetal bovine serum in the sample material
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: 6% cross-linked magnetized agarose
  • Average particle size: 25–30 μm
  • Suspension: 25% slurry
  • Ligand: CaptureSelect CH1-XL affinity ligand
  • Binding capacity: >4 mg IgG/mL slurry
  • Elution conditions: 50 mM sodium acetate, pH 4.0/0.1 M glycine, pH 3.0
Specifications
Binding Capacity>4 mg IgG/mL slurry
Ligand TypeCH1-XL affinity ligand
Matrix6% cross-linked magnetized agarose
Quantity1 mL
FormResin Slurry
Particle Size25 to 30 μm
Product LineCaptureSelect
TypeMagnetic Agarose Bead
Unit SizeEach