Precision™ Concentric Ring Electrical Steaming Baths
Thermo Scientific™

Precision™ Concentric Ring Electrical Steaming Baths

Thermo Scientific™ Precision™ concentric ring electrical steaming water baths have been discontinued.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)No. of PositionsElectrical Requirements
2896TS
also known as 2896
7.5 L4120 V 50/60 Hz
28977.5 L4230 V 50/60 Hz
289815 L8120 V 50/60 Hz
289915 L8230 V 50/60 Hz
Catalog number 2896TS
also known as 2896
Price (INR)
-
Capacity (Metric):
7.5 L
No. of Positions:
4
Electrical Requirements:
120 V 50/60 Hz

The Thermo Scientific™ Precision™ concentric ring electrical steaming water baths have been discontinued. For your water bath needs, please see our current offerings of Thermo Scientific water baths.

Baths accommodate various sizes of glassware and evaporating dishes, including round-bottom flasks. Flange-formed stainless-steel rings nest smoothly and become progressively larger as each concentric ring is removed. Copper-clad immersion heaters provide bath temperatures up to 100°C.

  • 4- and 8-hole models enlarge from 0.88in. (2.2cm) to maximum opening diameters of 5 and 6in. (12.7 and 15.2cm), respectively
  • Copper-clad immersion heaters provide bath temperature up to 100°C
  • Center ring cover is 1in.(2.5cm) in diameter
  • Easy to operate with a four-position switch and three different heat settings
  • Adjustable water level regulator maintains bath depth at the desired level
  • Stainless-steel cabinets with flanged cabinet top for tabletop installations
  • Rugged epoxy-coated, corrosion-resistant stand
  • Accommodate most laboratory glassware
  • Baths may also be flush-mounted in benchtop

Ordering Information:

Wiring of switch must be completed onsite.

Includes:

Precision ring sets, constant water level device (with 0.5in. inlet for water supply), adjustable drain tube, and welded angle-iron support frame with hot-rolled steel legs, fasteners and lockwashers

Requires:

Hardwire installation by a qualified electrician and hook-up to a continuous water source

Recommended for:

  • Copper strip corrosion tests
  • Melting agar
  • Metallurgical analysis
  • Thawing cryopreservation vials
  • Warranty varies by country, please contact us for details
Specifications
Amperage9.2 A
Diameter (English) Min. Opening0.88 in.
Diameter (Metric) Min. Opening122 mm
Frequency50/60 Hz
Heat Up Time63 min. to 100°C
Heater Settings (Low, Medium, High)275w, 550w, 1100w (Low, Medium, High)
Height (English) Exterior With Support13 in.
Height (English) Exterior Without Support4 in.
Height (Metric) Exterior With Support33 cm
Height (Metric) Exterior Without Support10.2 cm
Length (English) Exterior With Support15 in.
Length (English) Exterior Without Support15 in.
Length (Metric) Exterior With Support38 cm
Length (Metric) Exterior Without Support38 cm
MaterialStainless Steel
Min. Opening Diameter (All Rings Removed)4 at 125 mm (5 in.)
No. of Positions4
Temperature (Metric) Maximum100°C
Voltage120 V
Width (English) Exterior With Support15 in.
Width (English) Exterior Without Support15 in.
Width (Metric) Exterior With Support38 cm
Width (Metric) Exterior Without Support38 cm
Capacity (Metric)7.5 L
Electrical Requirements120 V 50/60 Hz
Unit SizeEach