Ac-dC Phosphoramidite, standard grade, serum vial bottle
Thermo Scientific™

Ac-dC Phosphoramidite, standard grade, serum vial bottle

Ac-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Acetyl (Ac) base protection, for useRead more
Have Questions?
Catalog NumberQuantity
29-1727-072 g
Catalog number 29-1727-07
Price (INR)
0.01
Each
Quantity:
2 g
Price (INR)
0.01
Each

Ac-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Acetyl (Ac) base protection, for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. By using Ac-dC, Bz-dA, and DMF-dG monomers, fast cleavage and deprotection can be effected by a 1:1 mixture of aqueous ammonium hydroxide and aqueous methylamine (or AMA) in 10 min. This product is supplied in a 60 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.

Features:

  • Full chemical name – 5'-O-(4,4'-Dimethoxytrityl)-N4-Acetyl-2'-deoxycytidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)-phosphoramidite
  • Molecular formula – C41H50N5O8P
  • Base – dC
  • Base Protection – Ac
  • Molecular weight – 771.8 g/mol
  • CAS number – 154110-40-4
  • HPLC Purity – ≥98%
  • 31P NMR Purity – ≥98%
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AKTA Synthesizers, Applied Biosystems Synthesizers
FormatPowder
GradeStandard
Nucleic Acid TypeDNA
Nucleoside TypeDeoxycytidine (dC)
Product TypeAc-dC CEP
Purity98%
Quantity2 g
Reagent FeaturesFast deprotection
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).