(2S,3S)-2-Amino-3-methoxybutanoic acid, 98%
(2S,3S)-2-Amino-3-methoxybutanoic acid, 98%
(2S,3S)-2-Amino-3-methoxybutanoic acid, 98%
Acros Organics

(2S,3S)-2-Amino-3-methoxybutanoic acid, 98%

CAS: 104195-80-4 | C5H11NO3 | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Packaging:
Glass bottle
Catalog number 290030010
also known as 29003
HSN 29225090
Price (INR)
137,295.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
137,295.00
Each
Chemical Identifiers
CAS104195-80-4
IUPAC Name(2S,3S)-2-azaniumyl-3-methoxybutanoate
Molecular FormulaC5H11NO3
InChI KeyFYCWLJLGIAUCCL-IMJSIDKUSA-N
SMILESCO[C@@H](C)[C@H]([NH3+])C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Specific optical rotation+25° to +29° (25°C, 589 nm) (C=0.5, 6 N HCl)
Titration with HClO4>=97.5 %
Infrared spectrumConforms
Appearance (Form)Powder or flakes
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only