4-tert-Butylcalix[4]arene, 99%
4-tert-Butylcalix[4]arene, 99%
4-tert-Butylcalix[4]arene, 99%
Acros Organics

4-tert-Butylcalix[4]arene, 99%

CAS: 60705-62-6 | C44H56O4 | 648.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 290220050
HSN 29071990
Price (INR)
9,398.00
Each
Quantity:
5 g
Request bulk or custom format
Price (INR)
9,398.00
Each
Chemical Identifiers
CAS60705-62-6
IUPAC Name5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol
Molecular FormulaC44H56O4
InChI KeyNVKLTRSBZLYZHK-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or flakes
Appearance (Color)White to light beige
Infrared spectrumConforms
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only