See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
Home
▸
Shop All Products
▸
Chromatography Supplies
▸
Chromatography Ferrule and Nut Kits
▸
Finger Tite Connectors for Thermo Scientific™ and Agilent™ Instruments
Actual product may vary
Finger Tite Connectors for Thermo Scientific™ and Agilent™ Instruments
Thermo Scientific Finger Tite Connectors for Thermo Scientific and Agilent Instruments offer easier fit, reliability and leak-free connections.
Have Questions?
Change view
Catalog Number
Type
290SA131
Split/Splitless and MS Starter Kit
290SA132
Split/Splitless and FID Starter Kit
290SA133
Split/Splitless Injector Base Seal
3 Options
Catalog number
290SA131
Price (INR)
58,024.00
Each
Add to cart
Type:
Split/Splitless and MS Starter Kit
Price (INR)
58,024.00
Each
Add to cart
Thermo Scientific™ Finger Tite Connectors for Thermo Scientific™ and Agilent Instruments offer easier fit, reliability and leak-free connections.
Advantages over conventional graphite/polymer ferrules:
Perfect metal-to-metal seal eliminates contamination and gives lower air/water background
No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures
Easy to handle with the nut touchable even with a hot injector/detector
No tools required
Specifications
Description
MS Starter Kit; 1/Pk.
For Use With (Equipment)
TRACE 1300 Series GC and Agilent GC Instruments
Type
Split/Splitless and MS Starter Kit
Quantity
1/Pk.