Search
Search

| Catalog Number | Material | Diameter (Metric) |
|---|---|---|
| 290VA193 | 15% Graphite, 85% Vespel | For 0.45 to 0.53 mm ID column |
| 290GA139 also known as 072635TH | 100% Graphite | For 0.1 to 0.32 mm ID column |
| 290GA140 | 100% Graphite | For 0.45 to 0.53 mm ID column |
| 290VA191 | 15% Graphite, 85% Vespel | For 0.1 to 0.25 mm ID column |
| 290VA192 | 15% Graphite, 85% Vespel | For 0.32 mm ID column |
Ordering Information
Packaged in contaminant-free, individual blister packs.