(Aminomethyl)phosphonic acid, 99%, Thermo Scientific Chemicals
(Aminomethyl)phosphonic acid, 99%, Thermo Scientific Chemicals
(Aminomethyl)phosphonic acid, 99%, Thermo Scientific Chemicals
Acros Organics

(Aminomethyl)phosphonic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
291640010
also known as 29164
1 gGlass bottle
Catalog number 291640010
also known as 29164
HSN 29319090
Price (INR)
12,712.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
12,712.00
Each
Chemical Identifiers
CAS1066-51-9
IUPAC Name(aminomethyl)phosphonic acid
Molecular FormulaCH6NO3P
InChI KeyMGRVRXRGTBOSHW-UHFFFAOYSA-N
SMILESNCP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.5 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only