3,4-Hexanedione, 96%
3,4-Hexanedione, 96%
3,4-Hexanedione, 96%
Acros Organics

3,4-Hexanedione, 96%

CAS: 4437-51-8 | C6H10O2 | 114.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
25 mL
Packaging:
Glass bottle
Catalog number 293272500
also known as 29327
HSN 29141990
Price (INR)
30,659.00
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
30,659.00
Each
Chemical Identifiers
CAS4437-51-8
IUPAC Namehexane-3,4-dione
Molecular FormulaC6H10O2
InChI KeyKVFQMAZOBTXCAZ-UHFFFAOYSA-N
SMILESCCC(=O)C(=O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Refractive index1.4090 to 1.4150 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=95 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only