3,5-Difluorobenzaldehyde, 98+%
3,5-Difluorobenzaldehyde, 98+%
3,5-Difluorobenzaldehyde, 98+%
Acros Organics

3,5-Difluorobenzaldehyde, 98+%

CAS: 32085-88-4 | C7H4F2O | 142.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 293470050
HSN 29130090
Price (INR)
11,623.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
11,623.00
Each
Chemical Identifiers
CAS32085-88-4
IUPAC Name3,5-difluorobenzaldehyde
Molecular FormulaC7H4F2O
InChI KeyASOFZHSTJHGQDT-UHFFFAOYSA-N
SMILESFC1=CC(C=O)=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.0 %
Appearance (Form)Liquid or solid
Appearance (Color)Clear colorless to light yellow to light green
Refractive index1.4920 to 1.4950 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only