CaptureSelect™ Factor II(a) Affinity Matrix
Thermo Scientific™

CaptureSelect™ Factor II(a) Affinity Matrix

CaptureSelect Factor II(a) Affinity Matrix is specifically engineered for the purification of human coagulation factors II (prothrombin) and IIa (thrombin) through recognition of a B-chain epitope found on both prothrombin and thrombin.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
294405201010 mL
29440520055 mL
294405205050 mL
Catalog number 2944052010
Price (INR)
230,585.00
Each
Quantity:
10 mL
Price (INR)
230,585.00
Each
CaptureSelect Factor II(a) Affinity Matrix is specifically engineered for the purification of human coagulation factors II (prothrombin) and IIa (thrombin) through recognition of a B-chain epitope found on both prothrombin and thrombin.

Due to its calcium-independent binding to the B-chain epitope, this resin is particularly well-suited for purifying native factors II and IIa from human plasma sources. Additionally, it provides high purity and yield in the purification of recombinant forms of these factors from clarified culture harvests.

Features of this affinity matrix include:

  • Highly selective for factors II (prothrombin) and IIa (thrombin), without cross reactivity to other human plasma proteins
  • Binds without the need for calcium
  • Enables efficient purification of prothrombin from plasma with high yields and retained activity
  • Mild elution at neutral pH using magnesium chloride
  • Excellent scalability
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (animal origin-free).

Main characteristics

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect Factor II(a) affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: > 6 mg/mL
  • Binding conditions: 50 mM Tris, 150 mM NaCl, pH 7
  • Elution conditions: 50 mM Tris, 2 M MgCl2 pH 7
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding Capacity>6 g/L at 5 min. residence time
Flow Rate50 to 200 cm/hr. (up to 2 bar)
FormatAgarose Bead
Formulation20% (v/v) ethanol
Ligand TypeCaptureSelect FactorII(a) Affinity Ligand
Quantity10 mL
TargetB-chain epitope on Factor II and IIa
FormResin Slurry
Particle Size65 ±10 μm
Product LineCaptureSelect
TypeAffinity Resin
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C. Protect from direct sunlight.