CaptureSelect™ EPO Affinity Matrix
Thermo Scientific™

CaptureSelect™ EPO Affinity Matrix

CaptureSelect EPO Affinity Matrix is designed for the purification of Erythropoietin (EPO) and/or Darbepoetin (DPO) from recombinant sources.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
29441020055 mL
294410201010 mL
294410205050 mL
Catalog number 2944102005
Price (INR)
99.00
Each
Quantity:
5 mL
Price (INR)
99.00
Each

CaptureSelect EPO Affinity Matrix is designed for the purification of Erythropoietin (EPO) and/or Darbepoetin (DPO) from recombinant sources.

Features of the CaptureSelect EPO Affinity Matrix include:

  • EPO purification with high purity and yield from recombinant sources
  • The CaptureSelect EPO affinity ligand recognizes a proteinaceous epitope thereby covering all EPO glycosylation isoforms present in the feed
  • Binding both EPO & DPO enabling purification of these molecules
  • Efficient elution at neutral pH (50 mM Tris 0.5 M CaCl2 pH 7.0) and acidic pH (20 mM Citric acid pH 3.0)
  • Excellent scalability
  • Non-animal-derived

The high selectivity and yields obtained using CaptureSelect EPO Affinity Matrix enable a robust, fast, and efficient purification process with excellent purity in one step, eliminating the need for multiple purification stages.

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created using a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Matrix: agarose-based
  • Average particle size: 65μm ± 10μm
  • Ligand: CaptureSelect EPO affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: > 6 mg EPO/mL resin
  • Binding conditions: PBS or TBS (Tris buffered saline) pH 7.4
  • Elution conditions: 50 mM Tris 0.5 M CaCl2 pH 7.0 (neutral), 20 mM citric acid, pH 3 (acidic)
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding Capacity>6 mg EPO/mL Resin
Flow Rate50 to 200 cm/hr. (up to 2 bar)
FormatAgarose bead
Formulation20% (v/v) ethanol
Quantity5 mL
Capacity (Metric)5 mL
FormResin Slurry
Particle Size63 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C. Protect from direct daylight.